Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
beat-IIIc
Entrez Gene ID
35037
Gene Name
beaten path IIIc
Gene Aliases
BcDNA:RE14414, Beat-IIIC, Beat-IIIc, CG15138, CG6365, Dmel\CG15138, Dmel_CG15138, beat IIIc
Chromosome Location
Chr.2L: 17187637 - 17260734 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032629
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135986.3 NP_609830.2 2 - 3 497 74
NM_165209.2 NP_724042.1 2 - 3 497 74
GenBank mRNA AY075458.1 - 2 - 3 74

View Full Product Details