Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sa
- Entrez Gene ID
-
40367
- Gene Name
- spermatocyte arrest
- Gene Aliases
-
CG11308, Dmel\CG11308, Dmel_CG11308, Sa, TAF-like, TAF8L, dTAF8L, gfb, ms(3)VO45, ms(3)VO5, ms(3)sa, tTAF
- Chromosome Location
-
Chr.3L: 21405425 - 21406738 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002842
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141054.3 |
NP_649311.1 |
1 - 2 |
159 |
66 |
GenBank mRNA |
AY752879.1 |
- |
1 - 2 |
196 |
66 |
View Full Product Details