Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Oda
- Entrez Gene ID
-
36307
- Gene Name
- Ornithine decarboxylase antizyme
- Gene Aliases
-
AZ, CG16747, D-AZ, Dmel\CG16747, Dmel_CG16747, ODA, guf, l(2)02833, l(2)k09037, oda
- Chromosome Location
-
Chr.2R: 12166295 - 12174264 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014184
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165864.3 |
NP_725104.1 |
1 - 2 |
360 |
95 |
View Full Product Details