Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SmD3
Entrez Gene ID
36306
Gene Name
Small ribonucleoprotein particle protein SmD3
Gene Aliases
CG8427, Dmel\CG8427, Dmel_CG8427, SYM10, Sm D3, dD3, guf, guf2, l(2)131/7, l(2)154/1, l(2)90/37, smD3, smd3, snRNP Sm D3
Chromosome Location
Chr.2R: 12168731 - 12170003 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023167
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165866.3 NP_725106.1 1 - 2 468 132
GenBank mRNA AF038598.1 - 1 - 2 473 132
AY113465.1 - 1 - 2 465 132
KX531492.1 - 1 - 2 322 132

View Full Product Details