Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- garz
- Entrez Gene ID
-
36337
- Gene Name
- gartenzwerg
- Gene Aliases
-
ARF1GEF, ArfGEF, CG 8487, CG8487, Dmel\CG8487, Dmel_CG8487, GBF1, GEF, Garz, ang
- Chromosome Location
-
Chr.2R: 12326360 - 12333860 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264560
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165881.2 |
NP_725133.1 |
8 - 9 |
5307 |
94 |
View Full Product Details