Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- nemy
- Entrez Gene ID
-
36395
- Gene Name
- no extended memory
- Gene Aliases
-
CG13147, CG8776, CytB561, Dmel\CG8776, Dmel_CG8776, Nemy, P153, anon-WO0140519.146
- Chromosome Location
-
Chr.2R: 12659497 - 12670219 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261673
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_165934.3 |
NP_725208.1 |
2 - 3 |
498 |
81 |
GenBank mRNA |
BT003222.1 |
- |
2 - 3 |
496 |
81 |
View Full Product Details