Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dh31-R
Entrez Gene ID
36475
Gene Name
Diuretic hormone 31 Receptor
Gene Aliases
CG17043, CG17415, CG32843, DH31, DH31-R, DH31-R1, DH31R, Dh31-R1, Dh31R, Dh31r, DmeCG32843, Dmel\CG32843, Dmel_CG32843, anon-WO0170980.136, anon-WO0170980.137, dh31-R1
Chromosome Location
Chr.2R: 13267798 - 13295927 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052843
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274021.2 NP_001260950.1 9 - 10 1849 60
NM_001274022.2 NP_001260951.1 9 - 10 1849 60
NM_165979.3 NP_725278.1 9 - 10 1849 60

View Full Product Details