The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057510.4 | NP_476858.1 | 2 - 3 | 418 | 80 |
NM_165994.2 | NP_725302.1 | 2 - 3 | 349 | 80 | |
NM_165995.2 | NP_725303.1 | 2 - 3 | 320 | 80 | |
NM_165996.2 | NP_725304.1 | 1 - 2 | 469 | 80 | |
NM_165998.2 | NP_725306.1 | 2 - 3 | 251 | 80 | |
GenBank mRNA | AY061142.1 | - | 2 - 3 | 393 | 80 |
L12446.1 | - | 2 - 3 | 305 | 80 |