The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057510.4 | NP_476858.1 | 5 - 6 | 817 | 74 |
NM_165994.2 | NP_725302.1 | 5 - 6 | 748 | 74 | |
NM_165995.2 | NP_725303.1 | 5 - 6 | 719 | 74 | |
NM_165996.2 | NP_725304.1 | 4 - 5 | 868 | 74 | |
NM_165998.2 | NP_725306.1 | 5 - 6 | 650 | 74 | |
GenBank mRNA | AY061142.1 | - | 5 - 6 | 792 | 74 |
L12446.1 | - | 5 - 6 | 704 | 74 |