Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Papss
Entrez Gene ID
40167
Gene Name
PAPS synthetase
Gene Aliases
CG8363, DmPAPSS, Dmel\CG8363, Dmel_CG8363, PAPS, PAPSS, Paps, l(3)76BDn, paps, papss
Chromosome Location
Chr.3L: 19845293 - 19860925 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020389
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043152.2 NP_001036617.1 3 - 4 681 69
NM_001275143.1 NP_001262072.1 3 - 4 384 69
NM_001275144.1 NP_001262073.1 3 - 4 638 69
NM_079447.4 NP_524171.2 2 - 3 348 69
NM_168821.3 NP_730457.1 3 - 4 369 69
NM_168822.3 NP_730458.1 3 - 4 346 69
NM_168823.3 NP_730459.1 2 - 3 452 69
NM_168824.3 NP_730460.1 2 - 3 409 69
GenBank mRNA AY051724.1 - 3 - 4 333 69
BT100002.1 - 2 - 3 345 69

View Full Product Details