Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Six4
Entrez Gene ID
40297
Gene Name
CG3871-PA
Gene Aliases
CG3871, D-Six4, D-six4, DSix4, Dmel\CG3871, Dmel_CG3871, Dsix4, six4, six4/5
Chromosome Location
Chr.3L: 20788886 - 20792777 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027364
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140999.3 NP_649256.1 1 - 2 652 63
NM_168872.1 NP_730566.1 1 - 2 229 63
GenBank mRNA AF099185.1 - 1 - 2 618 63
AF247709.1 - 1 - 2 63
BT028820.2 - 1 - 2 178 63
BT031315.1 - 1 - 2 643 63

View Full Product Details