Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
siz
Entrez Gene ID
40327
Gene Name
schizo
Gene Aliases
CG10577, CG12980, CG12981, CG32434, Dmel\CG32434, Dmel_CG32434, Loner, Schizo, loner
Chromosome Location
Chr.3L: 21034535 - 21075444 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026179
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_168881.3 NP_730594.1 1 - 2 764 72
GenBank mRNA AY375487.1 - 1 - 2 760 72
BT150471.1 - 1 - 2 760 72

View Full Product Details