Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldh-III
Entrez Gene ID
45398
Gene Name
Aldehyde dehydrogenase type III
Gene Aliases
ALDH, ALDH-III, AldhIII, CG11140, CT41571, Dhap, Dmel\CG11140, Dmel_CG11140, FALDH, l(2)03610
Chromosome Location
Chr.2R: 7466879 - 7477162 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010548
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273843.1 NP_001260772.1 5 - 6 676 67
NM_001273844.1 NP_001260773.1 5 - 6 803 67
NM_001273846.1 NP_001260775.1 5 - 6 676 67
NM_136441.4 NP_610285.5 7 - 8 1142 67
NM_165526.3 NP_724560.1 4 - 5 875 67
NM_165527.3 NP_724561.2 5 - 6 714 67
NM_165528.3 NP_724562.2 7 - 8 1663 67
NM_165529.3 NP_724563.2 5 - 6 622 67
NM_165530.3 NP_724564.2 5 - 6 803 67
NM_165531.3 NP_724565.3 7 - 8 1018 67
NM_165532.3 NP_724566.3 7 - 8 922 67
NM_206050.3 NP_995772.1 5 - 6 676 67
GenBank mRNA BT003248.1 - 4 - 5 884 67
BT003252.1 - 5 - 6 771 67
BT044325.1 - 5 - 6 675 67
BT126071.1 - 7 - 8 1169 67

View Full Product Details