Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldh-III
Entrez Gene ID
45398
Gene Name
Aldehyde dehydrogenase type III
Gene Aliases
ALDH, ALDH-III, AldhIII, CG11140, CT41571, Dhap, Dmel\CG11140, Dmel_CG11140, FALDH, l(2)03610
Chromosome Location
Chr.2R: 7466879 - 7477162 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010548
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273844.1 NP_001260773.1 9 - 10 1507 82
NM_001273846.1 NP_001260775.1 9 - 10 1380 82
NM_165528.3 NP_724562.2 11 - 12 2367 82
NM_165531.3 NP_724565.3 11 - 12 1722 82
NM_206050.3 NP_995772.1 9 - 10 1380 82
GenBank mRNA BT044325.1 - 9 - 10 1379 82

View Full Product Details