Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lig
Entrez Gene ID
35771
Gene Name
lingerer
Gene Aliases
CG8715, Dmel\CG8715, Dmel_CG8715
Chromosome Location
Chr.2R: 8067977 - 8078815 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020279
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014503.2 NP_001014503.1 7 - 8 2779 72
NM_001169610.2 NP_001163081.1 8 - 9 2772 72
NM_001169611.1 NP_001163082.1 8 - 9 2796 72
NM_001169612.2 NP_001163083.1 9 - 10 2838 72
NM_001259270.2 NP_001246199.1 8 - 9 2772 72
NM_001259271.2 NP_001246200.1 8 - 9 2772 72
NM_001273865.1 NP_001260794.1 7 - 8 2779 72
NM_001273866.2 NP_001260795.1 6 - 7 2559 72
NM_001316409.1 NP_001303338.1 6 - 7 2559 72
NM_136504.5 NP_610348.3 8 - 9 2772 72
NM_165586.5 NP_724643.2 8 - 9 2841 72
NM_206056.5 NP_995778.2 8 - 9 3127 72
GenBank mRNA AF276964.1 - 7 - 8 2779 72
AY069682.1 - 8 - 9 2796 72
BT100315.1 - 7 - 8 2745 72

View Full Product Details