Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lig
Entrez Gene ID
35771
Gene Name
lingerer
Gene Aliases
CG8715, Dmel\CG8715, Dmel_CG8715
Chromosome Location
Chr.2R: 8067977 - 8078815 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020279
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014503.2 NP_001014503.1 8 - 9 3308 92
NM_001169610.2 NP_001163081.1 9 - 10 3301 92
NM_001169611.1 NP_001163082.1 9 - 10 3325 92
NM_001169612.2 NP_001163083.1 10 - 11 3367 92
NM_001259270.2 NP_001246199.1 9 - 10 3301 92
NM_001273866.2 NP_001260795.1 7 - 8 3088 92
NM_001316409.1 NP_001303338.1 7 - 8 3088 92
NM_136504.5 NP_610348.3 9 - 10 3301 92
NM_165586.5 NP_724643.2 9 - 10 3370 92
NM_206056.5 NP_995778.2 9 - 10 3656 92
GenBank mRNA AF276964.1 - 8 - 9 3308 92
AY069682.1 - 9 - 10 3325 92

View Full Product Details