Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndg
Entrez Gene ID
36089
Gene Name
Nidogen/entactin
Gene Aliases
BEST:CK00552, CG12908, CT32053, Dmel\CG12908, Dmel_CG12908, ndg
Chromosome Location
Chr.2R: 10313891 - 10322104 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026403
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136731.2 NP_610575.1 11 - 12 4177 68
NM_206074.2 NP_995796.2 11 - 12 4177 68
GenBank mRNA BT031149.1 - 11 - 12 4150 68

View Full Product Details