Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lola
Entrez Gene ID
44548
Gene Name
longitudinals lacking
Gene Aliases
BEST:LD03274, BTB-IV, BcDNA:LD17006, BcDNA:RH31485, BtbIV, CG12052, CG18376, CG18378, CG18379, CG18380, CG18381, CG30012, CG30013, CG30014, Dmel\CG12052, Dmel_CG12052, LD03274, Lola, NEST:bs06a08, bs06a08.y1, btb, eyeful, fus7, l(2)00642, l(2)s3697, sw59
Chromosome Location
Chr.2R: 10481894 - 10543291 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283521
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032229.3 NP_001027400.1 3 - 4 972 55
NM_080027.6 NP_524766.2 3 - 4 1115 55
NM_170617.5 NP_724945.1 3 - 4 1115 55
NM_170618.5 NP_724946.1 3 - 4 1115 55
NM_170619.3 NP_724949.2 3 - 4 966 55
NM_170620.4 NP_724950.1 3 - 4 972 55
NM_170621.4 NP_724951.1 3 - 4 966 55
NM_170622.6 NP_724952.2 3 - 4 972 55
NM_170623.6 NP_724953.2 3 - 4 972 55
NM_176128.5 NP_788308.3 3 - 4 1176 55
NM_176129.5 NP_788309.2 3 - 4 1115 55
NM_176130.5 NP_788310.2 3 - 4 1115 55
NM_176131.5 NP_788311.2 3 - 4 1115 55
NM_176132.4 NP_788312.2 3 - 4 1176 55
NM_176133.5 NP_788313.1 3 - 4 972 55
NM_176134.4 NP_788314.1 3 - 4 972 55
NM_176135.4 NP_788315.1 3 - 4 966 55
NM_176136.5 NP_788316.2 3 - 4 972 55
NM_176138.5 NP_788318.1 3 - 4 972 55
NM_176139.3 NP_788319.1 3 - 4 972 55
NM_176140.4 NP_788320.1 3 - 4 972 55
NM_206082.4 NP_995804.1 3 - 4 1115 55
NM_206083.4 NP_995805.1 3 - 4 1115 55
NM_206084.4 NP_995806.1 3 - 4 1115 55
NM_206085.4 NP_995807.1 3 - 4 972 55
GenBank mRNA AB107272.1 - 3 - 4 1172 55
AB107274.1 - 3 - 4 1172 55
AB107277.1 - 3 - 4 1172 55
AB107278.1 - 3 - 4 1172 55
AB107279.1 - 3 - 4 1172 55
AB107280.1 - 3 - 4 1172 55
AB107281.1 - 3 - 4 1172 55
AB107282.1 - 3 - 4 1172 55
AB107284.1 - 3 - 4 1172 55
AB107285.1 - 3 - 4 1172 55
AB107286.1 - 3 - 4 1172 55
AB107287.1 - 3 - 4 1172 55
AB107288.1 - 3 - 4 1172 55
AB107292.1 - 3 - 4 1113 55
AB107294.1 - 3 - 4 1113 55
AB107297.1 - 3 - 4 1113 55
AB107298.1 - 3 - 4 1113 55
AB107299.1 - 3 - 4 1113 55
AB107300.1 - 3 - 4 1113 55
AB107301.1 - 3 - 4 1113 55
AB107302.1 - 3 - 4 1113 55
AB107304.1 - 3 - 4 1113 55
AB107305.1 - 3 - 4 1113 55
AB107306.1 - 3 - 4 1113 55
AB107307.1 - 3 - 4 1113 55
AB107308.1 - 3 - 4 1113 55
AB107312.1 - 3 - 4 969 55
AB107314.1 - 3 - 4 969 55
AB107317.1 - 3 - 4 969 55
AB107318.1 - 3 - 4 969 55
AB107319.1 - 3 - 4 969 55
AB107320.1 - 3 - 4 969 55
AB107321.1 - 3 - 4 969 55
AB107322.1 - 3 - 4 969 55
AB107324.1 - 3 - 4 969 55
AB107325.1 - 3 - 4 969 55
AB107326.1 - 3 - 4 969 55
AB107327.1 - 3 - 4 969 55
AB107328.1 - 3 - 4 969 55
AB107332.1 - 3 - 4 969 55
AB107334.1 - 3 - 4 969 55
AB107337.1 - 3 - 4 969 55
AB107338.1 - 3 - 4 969 55
AB107339.1 - 3 - 4 969 55
AB107340.1 - 3 - 4 969 55
AB107341.1 - 3 - 4 969 55
AB107342.1 - 3 - 4 969 55
AB107344.1 - 3 - 4 969 55
AB107345.1 - 3 - 4 969 55
AB107346.1 - 3 - 4 969 55
AB107347.1 - 3 - 4 969 55
AB107348.1 - 3 - 4 969 55
AY058586.1 - 3 - 4 956 55
BT004909.1 - 3 - 4 1102 55
BT120030.1 - 3 - 4 945 55

View Full Product Details