Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syx6
Entrez Gene ID
40373
Gene Name
Syntaxin 6
Gene Aliases
CG7736, CT23545, Dm Syx6, DmSyx6, Dmel\CG7736, Dmel_CG7736, SYN6, SYX6/10, Syx
Chromosome Location
Chr.2R: 11219539 - 11222480 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037084
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206092.3 NP_995814.1 2 - 3 579 79
NM_206093.3 NP_995815.1 2 - 3 328 79
NM_206094.3 NP_995816.1 1 - 2 684 79
GenBank mRNA AY089525.1 - 1 - 2 636 79
BT001663.1 - 2 - 3 577 79
BT003505.1 - 2 - 3 326 79

View Full Product Details