Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chn
Entrez Gene ID
36678
Gene Name
charlatan
Gene Aliases
CG11798, Chn, Dmel\CG11798, Dmel_CG11798, EPIL#6, EPIL6, anon-EST:Liang-1.13, chl, clone 1.13, l(2)02064, l(2)k04218, taco
Chromosome Location
Chr.2R: 15115236 - 15144783 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015371
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043081.2 NP_001036546.1 3 - 4 1039 123
NM_001043082.2 NP_001036547.1 3 - 4 1264 123
NM_001259401.2 NP_001246330.1 3 - 4 1171 123
NM_001259402.2 NP_001246331.1 3 - 4 1039 123
NM_001259403.2 NP_001246332.1 3 - 4 1171 123
NM_001259404.2 NP_001246333.1 3 - 4 1171 123
NM_001259405.2 NP_001246334.1 3 - 4 1039 123
NM_137169.4 NP_611013.1 3 - 4 1171 123
NM_206119.2 NP_995841.1 3 - 4 1264 123
GenBank mRNA BT021440.1 - 3 - 4 1245 123

View Full Product Details