Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fs
- Entrez Gene ID
-
2768836
- Gene Name
- Follistatin
- Gene Aliases
-
CG12955, CG12956, CG33466, Dmel\CG33466, Dmel_CG33466, Fol1, dFS, dFol1, dfs, fs
- Chromosome Location
-
Chr.2R: 15223560 - 15241589 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259878
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144119.3 |
NP_652376.2 |
5 - 6 |
2366 |
60 |
GenBank mRNA |
BT003621.1 |
- |
5 - 6 |
2356 |
60 |
View Full Product Details