Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlf
Entrez Gene ID
36750
Gene Name
Myelodysplasia/myeloid leukemia factor
Gene Aliases
BcDNA:SD18148, CG8295, Dmel\CG8295, Dmel_CG8295, MLF, dMLF, dmlf, mlf
Chromosome Location
Chr.2R: 15935893 - 15940073 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034051
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_166122.4 NP_725501.2 4 - 5 1096 88
NM_206125.2 NP_995847.1 4 - 5 1119 88

View Full Product Details