Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CheB53b
Entrez Gene ID
3346204
Gene Name
Chemosensory protein B 53b
Gene Aliases
CG33457, CG33524, Dmel\CG33524, Dmel_CG33524
Chromosome Location
Chr.2R: 16468209 - 16468934 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261293
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014531.1 NP_001014531.1 2 - 3 442 130

View Full Product Details