The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057775.4 | NP_477123.1 | 6 - 7 | 2823 | 148 |
NM_166200.3 | NP_725623.2 | 5 - 6 | 2902 | 148 | |
NM_206140.2 | NP_995862.1 | 6 - 7 | 2826 | 148 | |
GenBank mRNA | BT031264.1 | - | 6 - 7 | 2790 | 148 |