Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGAP54D
Entrez Gene ID
36996
Gene Name
Rho GTPase activating protein at 54D
Gene Aliases
CG6477, Dmel\CG6477, Dmel_CG6477
Chromosome Location
Chr.2R: 17586425 - 17590884 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034249
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274131.2 NP_001261060.1 2 - 3 717 74
NM_137392.3 NP_611236.2 2 - 3 913 74
GenBank mRNA BT133065.1 - 2 - 3 701 74

View Full Product Details