Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sema1b
Entrez Gene ID
37007
Gene Name
Semaphorin 1b
Gene Aliases
BcDNA:GH03186, CG6446, D-semaIII, Dmel\CG6446, Dmel_CG6446, Sema 1b, Sema III, Sema-1b, sema-III, sema1b
Chromosome Location
Chr.2R: 17672839 - 17683499 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016059
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169707.2 NP_001163178.1 6 - 7 2686 78
NM_137400.4 NP_611244.1 6 - 7 2597 78
NM_206157.2 NP_995879.1 6 - 7 2590 78
GenBank mRNA AF181625.1 - 6 - 7 2580 78
BT083389.1 - 6 - 7 2673 78

View Full Product Details