Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ktub
Entrez Gene ID
37400
Gene Name
king tubby
Gene Aliases
87D3S, CG9398, D-tulp, Dmel\CG9398, Dmel_CG9398, Dtulp, ESTS:87D3S, King-Tubby, King-tubby, TULP, Tulp, dTULP, dTulp, king-tubby, tulp
Chromosome Location
Chr.2R: 21101928 - 21123704 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015721
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137705.3 NP_611549.2 5 - 6 1784 57
NM_206189.2 NP_995911.1 5 - 6 1757 57
GenBank mRNA BT003201.1 - 5 - 6 1755 57

View Full Product Details