Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bw
Entrez Gene ID
37724
Gene Name
brown
Gene Aliases
CG17632, Dmel\CG17632, Dmel_CG17632, Pm, Su(w[co2]), Su(w[coJ]), ptm
Chromosome Location
Chr.2R: 23527805 - 23538499 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000241
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299840.1 NP_001286769.1 5 - 6 1438 91
NM_079100.3 NP_523824.1 5 - 6 1438 91
GenBank mRNA M20630.1 - 5 - 6 1444 91

View Full Product Details