Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slik
Entrez Gene ID
37893
Gene Name
Sterile20-like kinase
Gene Aliases
CG4527, DPlkk1, Dmel\CG4527, Dmel_CG4527, PLKK1, Plkk/Slik, Plkk1, SLIK, SLK, Slik1, Slk, dPlkk, dPlkk/Slik, slik, slk
Chromosome Location
Chr.2R: 24392129 - 24402680 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035001
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014549.3 NP_001014549.1 5 - 6 1220 99
NM_001144280.2 NP_001137752.1 5 - 6 1179 99
NM_138064.4 NP_611908.2 5 - 6 1220 99
NM_166669.4 NP_726441.1 5 - 6 1220 99
NM_206213.3 NP_995935.1 5 - 6 1613 99
NM_206214.4 NP_995936.2 5 - 6 1220 99
GenBank mRNA BT150436.1 - 5 - 6 1228 99

View Full Product Details