Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Start1
Entrez Gene ID
37927
Gene Name
CG3522-PA
Gene Aliases
CG3522, Dmel\CG3522, Dmel_CG3522, StART1, Start, start1
Chromosome Location
Chr.2R: 24574559 - 24577676 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035028
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259566.2 NP_001246495.1 11 - 12 2032 90
NM_001299919.1 NP_001286848.1 5 - 6 965 90
NM_138092.3 NP_611936.2 11 - 12 1865 90
NM_206217.3 NP_995939.1 11 - 12 2055 90
GenBank mRNA AY102683.1 - 11 - 12 2020 90
AY455866.1 - 11 - 12 2014 90

View Full Product Details