Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dhc62B
- Entrez Gene ID
-
38226
- Gene Name
- Dynein heavy chain at 62B
- Gene Aliases
-
CG15804, CG18613, DHC, DHC7C, Dhc, Dmel\CG15804, Dmel_CG15804, dhc
- Chromosome Location
-
Chr.3L: 1759172 - 1771758 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013811
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206236.3 |
NP_995958.2 |
6 - 7 |
6674 |
65 |
View Full Product Details