Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zormin
Entrez Gene ID
2769001
Gene Name
CG33484-PE
Gene Aliases
BcDNA:GH09541, CG11850, CG11952, CG11953, CG1282, CG32307, CG32309, CG32310, CG32311, CG33484, CG5699, CT36990, Dmel\CG33484, Dmel_CG33484
Chromosome Location
Chr.3L: 2117466 - 2151700 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052311
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043112.2 NP_001036577.2 3 - 4 753 69
NM_001043113.2 NP_001036578.2 3 - 4 753 69
NM_001043114.2 NP_001036579.2 3 - 4 753 69
NM_001274386.1 NP_001261315.1 3 - 4 753 69
NM_001274388.1 NP_001261317.1 3 - 4 753 69
NM_206240.2 NP_995962.2 3 - 4 753 69

View Full Product Details