Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sif
Entrez Gene ID
43892
Gene Name
still life
Gene Aliases
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
Chromosome Location
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104046.4 NP_001097516.3 17 - 18 6570 66
NM_001104047.5 NP_001097517.1 28 - 29 6303 66
NM_001104048.2 NP_001097518.2 30 - 31 5990 66
NM_001104049.4 NP_001097519.2 29 - 30 5758 66
NM_001274517.3 NP_001261446.1 29 - 30 6360 66
NM_001274518.2 NP_001261447.1 16 - 17 3258 66
NM_001274519.3 NP_001261448.1 29 - 30 6360 66
NM_001300018.2 NP_001286947.1 28 - 29 7342 66
NM_001347763.1 NP_001334674.1 30 - 31 6306 66
NM_001347764.1 NP_001334675.1 30 - 31 6315 66
NM_079908.3 NP_524647.2 17 - 18 6543 66
NM_168126.4 NP_729084.1 28 - 29 5313 66
NM_206278.2 NP_996000.1 17 - 18 6570 66
GenBank mRNA AY118305.1 - 17 - 18 6548 66

Gene Symbol
CG46320
Entrez Gene ID
30522409
Gene Name
CG46320-PA
Gene Aliases
Dmel\CG46320, Dmel_CG46320, sif-5'utr
Chromosome Location
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347801.1 NP_001334709.1 28 - 29 6303 66
NM_001347802.1 NP_001334710.1 30 - 31 6306 66

View Full Product Details