Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sif
Entrez Gene ID
43892
Gene Name
still life
Gene Aliases
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
Chromosome Location
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104046.4 NP_001097516.3 18 - 19 6734 63
NM_001104047.5 NP_001097517.1 29 - 30 6467 63
NM_001104048.2 NP_001097518.2 31 - 32 6154 63
NM_001104049.4 NP_001097519.2 30 - 31 5922 63
NM_001274517.3 NP_001261446.1 30 - 31 6524 63
NM_001274518.2 NP_001261447.1 17 - 18 3422 63
NM_001274519.3 NP_001261448.1 30 - 31 6524 63
NM_001300018.2 NP_001286947.1 29 - 30 7506 63
NM_001347763.1 NP_001334674.1 31 - 32 6470 63
NM_001347764.1 NP_001334675.1 31 - 32 6479 63
NM_079908.3 NP_524647.2 18 - 19 6707 63
NM_168126.4 NP_729084.1 29 - 30 5477 63
NM_206278.2 NP_996000.1 18 - 19 6734 63
GenBank mRNA AY118305.1 - 18 - 19 6712 63

Gene Symbol
CG46320
Entrez Gene ID
30522409
Gene Name
CG46320-PA
Gene Aliases
Dmel\CG46320, Dmel_CG46320, sif-5'utr
Chromosome Location
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347801.1 NP_001334709.1 29 - 30 6467 63
NM_001347802.1 NP_001334710.1 31 - 32 6470 63

View Full Product Details