Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sif
Entrez Gene ID
43892
Gene Name
still life
Gene Aliases
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
Chromosome Location
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104046.4 NP_001097516.3 19 - 20 7106 58
NM_001104047.5 NP_001097517.1 30 - 31 6839 58
NM_001104048.2 NP_001097518.2 32 - 33 6526 58
NM_001104049.4 NP_001097519.2 31 - 32 6294 58
NM_001274517.3 NP_001261446.1 31 - 32 6896 58
NM_001274518.2 NP_001261447.1 18 - 19 3794 58
NM_001274519.3 NP_001261448.1 31 - 32 6896 58
NM_001300018.2 NP_001286947.1 30 - 31 7878 58
NM_001347763.1 NP_001334674.1 32 - 33 6842 58
NM_001347764.1 NP_001334675.1 32 - 33 6851 58
NM_079908.3 NP_524647.2 19 - 20 7079 58
NM_168126.4 NP_729084.1 30 - 31 5849 58
NM_206278.2 NP_996000.1 19 - 20 7106 58
GenBank mRNA AY118305.1 - 19 - 20 7084 58

Gene Symbol
CG46320
Entrez Gene ID
30522409
Gene Name
CG46320-PA
Gene Aliases
Dmel\CG46320, Dmel_CG46320, sif-5'utr
Chromosome Location
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347801.1 NP_001334709.1 30 - 31 6839 58
NM_001347802.1 NP_001334710.1 32 - 33 6842 58

View Full Product Details