Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smid
Entrez Gene ID
38824
Gene Name
smallminded
Gene Aliases
CG8571, Dmel\CG8571, Dmel_CG8571, Smid, l(3)j6B8, l(3)j8A6, l(3)s2898
Chromosome Location
Chr.3L: 7381614 - 7385637 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016983
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169900.2 NP_001163371.1 4 - 5 682 55
NM_001274583.1 NP_001261512.1 4 - 5 828 55
NM_079235.4 NP_523959.2 4 - 5 685 55
NM_206287.3 NP_996009.2 4 - 5 595 55
GenBank mRNA BT124778.1 - 4 - 5 826 55
X99207.1 - - 646 55

View Full Product Details