Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lqf
Entrez Gene ID
38846
Gene Name
liquid facets
Gene Aliases
0110/27, CG8532, Dmel\CG8532, Dmel_CG8532, E(faf)bE25, Epsin, LqF, Lqf, anon-WO0118547.239, dEpsin, epsin, l(3)S011027
Chromosome Location
Chr.3L: 7528780 - 7540012 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104059.2 NP_001097529.1 5 - 6 1935 62
NM_001274596.1 NP_001261525.1 5 - 6 1923 62
NM_001274597.1 NP_001261526.1 5 - 6 1923 62
NM_001274598.1 NP_001261527.1 6 - 7 2080 62
NM_001274599.1 NP_001261528.1 6 - 7 2080 62
NM_001274600.1 NP_001261529.1 4 - 5 1745 62
NM_001274601.1 NP_001261530.1 4 - 5 1745 62
NM_079238.3 NP_523962.2 5 - 6 1923 62
NM_168219.1 NP_729266.2 6 - 7 1858 62
NM_168220.1 NP_729267.1 4 - 5 1745 62
NM_168221.3 NP_729268.2 5 - 6 1923 62
NM_206291.2 NP_996013.1 6 - 7 2080 62
GenBank mRNA BT133163.1 - 6 - 7 2066 62

View Full Product Details