Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGRP-LA
Entrez Gene ID
39062
Gene Name
Peptidoglycan recognition protein LA
Gene Aliases
CG18614, CG32042, CG4361, CG4384, Dmel\CG32042, Dmel_CG32042, PGRPLA, pgrp-la
Chromosome Location
Chr.3L: 9334332 - 9338336 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035975
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274694.1 NP_001261623.1 3 - 4 532 64
NM_206304.2 NP_996026.1 4 - 5 885 64
NM_206305.2 NP_996027.1 4 - 5 873 64
NM_206306.2 NP_996028.1 3 - 4 780 64
NM_206307.2 NP_996029.1 3 - 4 793 64
GenBank mRNA AF207535.1 - 3 - 4 765 64
AF207536.1 - 3 - 4 781 64
AY060314.1 - 4 - 5 872 64
BT001411.1 - 3 - 4 764 64

View Full Product Details