Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-52
Entrez Gene ID
31499
Gene Name
CG15929-PA
Gene Aliases
CG15929, Dmel\CG15929, Dmel_CG15929, Lin-52, Lin52, dLIN-52, dLin-52, dLin52, dlin-52, dlin52
Chromosome Location
Chr.X: 5753390 - 5754958 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029800
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297979.1 NP_001284908.1 1 - 2 107 64
NM_132028.3 NP_572256.1 1 - 2 107 64
GenBank mRNA AY788901.1 - 1 - 2 33 64

View Full Product Details