Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GAA1
- Entrez Gene ID
-
31517
- Gene Name
- Glycosylphosphatidylinositol anchor attachment 1
- Gene Aliases
-
CG3033, Dmel\CG3033, Dmel_CG3033
- Chromosome Location
-
Chr.X: 5870841 - 5873357 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029818
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132045.4 |
NP_572273.2 |
2 - 3 |
1958 |
65 |
View Full Product Details