Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCTS1
Entrez Gene ID
31536
Gene Name
Malignant T cell amplified sequence 1
Gene Aliases
CG5941, Dmel\CG5941, Dmel_CG5941, MCT-1, anon-WO03040301.150, anon-WO03040301.151
Chromosome Location
Chr.X: 6037892 - 6039576 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029833
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258612.2 NP_001245541.1 2 - 3 244 68
NM_132060.4 NP_572288.1 2 - 3 244 68
GenBank mRNA AY051886.1 - 2 - 3 217 68

View Full Product Details