Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
raptor
Entrez Gene ID
31543
Gene Name
CG4320-PB
Gene Aliases
4320, CG4320, Dmel\CG4320, Dmel_CG4320, Raptor, dRAPTOR, dRap, dRaptor, raptor/CG4320
Chromosome Location
Chr.X: 6082634 - 6090201 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029840
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297995.1 NP_001284924.1 1 - 2 1931 56
NM_132066.2 NP_572294.2 1 - 2 1931 56

View Full Product Details