Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CadN2
Entrez Gene ID
35071
Gene Name
Cadherin-N2
Gene Aliases
CG42829, CG7527, CT23007, DN-Cad2, DN-cad2, DN-like, DNcad2, DmNCad2, Dmel\CG42829, Dmel_CG42829, N-Cad2, NCad2, Ncad2, cadN2
Chromosome Location
Chr.2L: 17786971 - 17875247 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262018
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042903.4 NP_001036368.2 8 - 9 2430 75
NM_136011.4 NP_609855.3 9 - 10 3124 75
GenBank mRNA BT132666.1 - 8 - 9 2397 75

View Full Product Details