Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Grip71
- Entrez Gene ID
-
35130
- Gene Name
- CG10346-PA
- Gene Aliases
-
CG10346, Dgp71WD, Dgrip71, Dmel\CG10346, Dmel_CG10346, Grip71wd
- Chromosome Location
-
Chr.2L: 18708846 - 18711297 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032705
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136058.5 |
NP_609902.3 |
2 - 3 |
577 |
72 |
View Full Product Details