The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080273.3 | NP_525012.2 | 4 - 5 | 565 | 72 |
NM_144389.2 | NP_652646.1 | 4 - 5 | 560 | 72 | |
NM_165502.2 | NP_724524.1 | 4 - 5 | 554 | 72 | |
GenBank mRNA | AF274014.1 | - | 4 - 5 | 552 | 72 |
AY069049.1 | - | 4 - 5 | 552 | 72 |