Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpn13
Entrez Gene ID
36545
Gene Name
Regulatory particle non-ATPase 13
Gene Aliases
CG13349, Dmel\CG13349, Dmel_CG13349, GC13349, p42E, rpn13
Chromosome Location
Chr.2R: 13958545 - 13960404 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033886
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144192.2 NP_001137664.1 2 - 3 837 63
NM_137073.3 NP_610917.1 3 - 4 824 63
NM_166025.2 NP_725346.1 3 - 4 837 63
GenBank mRNA AY061027.1 - 2 - 3 837 63

View Full Product Details