Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cp1
Entrez Gene ID
36546
Gene Name
Cysteine proteinase-1
Gene Aliases
CATL_DROME, CG6692, CL, CP1, CT20780, CTSL, CatL, Cath L, CathL, CysP-1, DCP-1, DCP1, Dcp-1, Dcp1, Dmel\CG6692, Dmel_CG6692, cp1, fs(2)50Ca
Chromosome Location
Chr.2R: 13960675 - 13967981 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013770
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079011.3 NP_523735.2 2 - 3 204 64
NM_166026.3 NP_725347.1 2 - 3 321 64
GenBank mRNA BT016071.1 - 2 - 3 332 64
U75652.1 - 2 - 3 248 64

View Full Product Details