Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RN-tre
Entrez Gene ID
36554
Gene Name
Related to the N terminus of tre oncogene
Gene Aliases
CG13351, CG8085, DRN-tre, Dmel\CG8085, Dmel_CG8085
Chromosome Location
Chr.2R: 13984815 - 13990707 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020620
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201996.3 NP_001188925.2 5 - 6 1492 74
NM_001274032.2 NP_001260961.1 6 - 7 1678 74
NM_166028.4 NP_725351.1 6 - 7 1678 74
GenBank mRNA AY061446.1 - 6 - 7 1642 74

View Full Product Details