Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rcd1
Entrez Gene ID
36560
Gene Name
Reduction in Cnn dots 1
Gene Aliases
CG8233, Dmel\CG8233, Dmel_CG8233, NSL3, Nsl3, cg8233
Chromosome Location
Chr.2R: 14148263 - 14153123 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033897
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274033.2 NP_001260962.1 7 - 8 2583 70
NM_001274034.2 NP_001260963.1 7 - 8 2950 70
NM_001274035.2 NP_001260964.1 6 - 7 2523 70
NM_001274036.2 NP_001260965.1 6 - 7 2890 70
NM_001299475.1 NP_001286404.1 8 - 9 2881 70
NM_137083.4 NP_610927.3 6 - 7 2755 70
NM_166031.3 NP_725357.2 6 - 7 2388 70
NM_166032.3 NP_725358.1 6 - 7 2225 70
GenBank mRNA AY069502.1 - 6 - 7 2730 70
BT100232.1 - 6 - 7 2354 70

View Full Product Details