Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aPKC
Entrez Gene ID
47594
Gene Name
atypical protein kinase C
Gene Aliases
CG10261, CG30475, CG42783, DaPKC, Dmel\CG42783, Dmel_CG42783, PKCl, PKCzeta, a-PKC, aPKC zeta, aPKC-zeta, aPKCzeta, aPkc, alphaPKCzeta, apkc, daPKC, dapkc, i, l(2)k06403, pkc-3, psu
Chromosome Location
Chr.2R: 14944458 - 14962969 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261854
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202010.4 NP_001188939.1 2 - 3 785 58

View Full Product Details